accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCS

CAS RN 622-59-3

IUPAC Name: 1-isothiocyanato-4-methylbenzene
Synonyms: 4-methylphenylisothiocyanate
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: ABQKHKWXTUVKGF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N=C=S
Boiling Point 
  • (3)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)