accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62217-35-0

IUPAC Name: dimethyl (2H-1,3-benzodithiol-2-yl)phosphonate
Synonyms: dimethyl 2H-1,3-benzodithiol-2-ylphosphonate
Molecular Weight (g/mol): 262.28
Molecular Formula: C9H11O3PS2
InChi Key: VKVOUPYVLWQXLB-UHFFFAOYSA-N
SMILES: COP(=O)(OC)C1SC2=CC=CC=C2S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results