accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3O

CAS RN 6222-17-9

IUPAC Name: 6-(acetyloxy)hexyl acetate
Synonyms: 1,6-hexanediol, diacetate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: ZMFWEWMHABZQNB-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results