accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6222-35-1

IUPAC Name: cyclohexyl propanoate
Synonyms: propionic acid cyclohexyl ester
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: MAMMVUWCKMOLSG-UHFFFAOYSA-N
SMILES: CCC(=O)OC1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results