accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62234-36-0

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 338.36
Molecular Formula: C16H22N2O6
InChi Key: WJKGPJRAGHSOLM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)