accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaNaOSOONNHNNOSOOOSOOOSOOO(E/Z)(Z)

CAS RN 6226-79-5

IUPAC Name: tetrasodium (4Z)-3-oxo-4-(2-{2-sulfonato-5-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: ponceau S
Molecular Weight (g/mol): 760.55
Molecular Formula: C22H12N4Na4O13S4
InChi Key: LZYGEJPERQQCIY-STNZDNLRSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(C(N\N=C2/C(=O)C(=CC3=CC(=CC=C23)S([O-])(=O)=O)S([O-])(=O)=O)=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Color 
  • (3)
  • (3)
  • (1)
Grade 
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (8)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)