accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClN

CAS RN 623-03-0

IUPAC Name: 4-chlorobenzonitrile
Synonyms: benzonitrile, 4-chloro-
Molecular Weight (g/mol): 137.57
Molecular Formula: C7H4ClN
InChi Key: GJNGXPDXRVXSEH-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
  • (2)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (1)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (7)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)