accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 623-17-6

IUPAC Name: (furan-2-yl)methyl acetate
Synonyms: 2-furanmethanol, acetate
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: CKOYRRWBOKMNRG-UHFFFAOYSA-N
SMILES: CC(=O)OCC1=CC=CO1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)