accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSH

CAS RN 623-51-8

IUPAC Name: ethyl 2-sulfanylacetate
Synonyms: ethyl thioglycolate
Molecular Weight (g/mol): 120.17
Molecular Formula: C4H8O2S
InChi Key: PVBRSNZAOAJRKO-UHFFFAOYSA-N
SMILES: CCOC(=O)CS
Boiling Point 
  • (3)
  • (2)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (3)
  • (5)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)