accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 623-57-4

IUPAC Name: 3-(dimethylamino)propane-1,2-diol
Synonyms: 3-(dimethylamino)propane-1,2-diol
Molecular Weight (g/mol): 119.16
Molecular Formula: C5H13NO2
InChi Key: QCMHUGYTOGXZIW-UHFFFAOYNA-N
SMILES: CN(C)CC(O)CO
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)