accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3COOCH3O

CAS RN 623-84-7

IUPAC Name: 1-(acetyloxy)propan-2-yl acetate
Synonyms: propylene glycol diacetate 1,2-propylene diacetate propylene acetate propyleneglycol diacetate methylethylene acetate methylethylene diacetate 1,2-diacetate 1,2-propanediol 1,2-propanediol, 1,2-diacetate 1,2-propylene glycol diacetate propylene diacetate
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: MLHOXUWWKVQEJB-UHFFFAOYNA-N
SMILES: CC(COC(C)=O)OC(C)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)