accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62345-76-0

IUPAC Name: 4-[2-(dimethylamino)ethoxy]aniline
Synonyms: 4-[2-(dimethylamino)ethoxy]aniline
Molecular Weight (g/mol): 180.25
Molecular Formula: C10H16N2O
InChi Key: CCCVQPGAXZNTIL-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results