accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 623564-18-1

IUPAC Name: ethyl 5H,6H,7H,8H-imidazo[1,2-a]pyrazine-2-carboxylate dihydrochloride
Synonyms: ethyl 5H,6H,7H,8H-imidazo[1,2-a]pyrazine-2-carboxylate dihydrochloride
Molecular Weight (g/mol): 268.14
Molecular Formula: C9H15Cl2N3O2
InChi Key: UVYMXIAFTVXRIT-UHFFFAOYSA-N
SMILES: Cl.Cl.CCOC(=O)C1=CN2CCNCC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results