accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 62373-80-2

IUPAC Name: 3-(4-methoxyphenoxy)benzaldehyde
Synonyms: 3-(4-methoxyphenoxy)benzaldehyde
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: WLFDEVVCXPTAQA-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC2=CC=CC(C=O)=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results