accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 624-18-0

IUPAC Name: dihydrogen benzene-1,4-diamine dichloride
Synonyms: P-phenylenediamine hcl
Molecular Weight (g/mol): 181.06
Molecular Formula: C6H10Cl2N2
InChi Key: IYXMNTLBLQNMLM-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)C=C1
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)