accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 624-31-7

IUPAC Name: 1-iodo-4-methylbenzene
Synonyms: benzene, 1-iodo-4-methyl-
Molecular Weight (g/mol): 218.04
Molecular Formula: C7H7I
InChi Key: UDHAWRUAECEBHC-UHFFFAOYSA-N
SMILES: CC1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (2)
  • (6)
  • (2)
Quantity 
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (8)
  • (2)
  • (6)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)