accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3C(Z)

CAS RN 624-48-6

IUPAC Name: 1,4-dimethyl (2Z)-but-2-enedioate
Synonyms: dimethyl maleate sipomer dmm dimethylester kyseliny maleinove
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: LDCRTTXIJACKKU-ARJAWSKDSA-N
SMILES: COC(=O)\C=C/C(=O)OC
Boiling Point 
  • (4)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (2)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (7)
  • (4)
  • (2)
  • (3)
  • (2)
  • (6)
  • (5)
  • (2)
  • (9)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)