accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 624-66-8

IUPAC Name: 1-iodoprop-1-yne
Synonyms: 1-iodoprop-1-yne
Molecular Weight (g/mol): 165.96
Molecular Formula: C3H3I
InChi Key: JHUPGXNKUPOSIE-UHFFFAOYSA-N
SMILES: CC#CI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results