accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 624-83-9

IUPAC Name: isocyanatomethane
Synonyms: methyl isocyanate isocyanate de methyle MIC methylcarbylamine
Molecular Weight (g/mol): 57.05
Molecular Formula: C2H3NO
InChi Key: HAMGRBXTJNITHG-UHFFFAOYSA-N
SMILES: CN=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results