CAS RN 6243-10-3
IUPAC Name:
cyclohexyl hexanoate
Synonyms:
cyclohexyl hexanoate
Molecular Weight (g/mol):
198.31
Molecular Formula:
C12H22O2
InChi Key:
DTNOERNOMHQUCN-UHFFFAOYSA-N
SMILES:
CCCCCC(=O)OC1CCCCC1
1
–
1
of
1
results
1