accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 62462-05-9

IUPAC Name: methyl 5-methoxy-3-oxopentanoate
Synonyms: methyl 5-methoxy-3-oxopentanoate
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: OPURNNHYTATPKM-UHFFFAOYSA-N
SMILES: COCCC(=O)CC(=O)OC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results