accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 625-22-9

IUPAC Name: dibutyl sulfate
Synonyms: sulfuric acid, dibutyl ester
Molecular Weight (g/mol): 210.29
Molecular Formula: C8H18O4S
InChi Key: LMEDOLJKVASKTP-UHFFFAOYSA-N
SMILES: CCCCOS(=O)(=O)OCCCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)