accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 625-27-4

IUPAC Name: 2-methylpent-2-ene
Synonyms: 2-methyl-2-pentene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: JMMZCWZIJXAGKW-UHFFFAOYSA-N
SMILES: CCC=C(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)