accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 625-60-5

IUPAC Name: 1-(ethylsulfanyl)ethan-1-one
Synonyms: ethanethioic acid, ethyl ester
Molecular Weight (g/mol): 104.17
Molecular Formula: C4H8OS
InChi Key: APTGPWJUOYMUCE-UHFFFAOYSA-N
SMILES: CCSC(C)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)