accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHCH3

CAS RN 625-84-3

IUPAC Name: 2,5-dimethyl-1H-pyrrole
Synonyms: 2,5-dimethylpyrrole
Molecular Weight (g/mol): 95.15
Molecular Formula: C6H9N
InChi Key: PAPNRQCYSFBWDI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N1
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)