accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3

CAS RN 625-86-5

IUPAC Name: 2,5-dimethylfuran
Synonyms: 2,5-dimethylfuran
Molecular Weight (g/mol): 96.13
Molecular Formula: C6H8O
InChi Key: GSNUFIFRDBKVIE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)O1
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (5)
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (8)