accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 626-15-3

IUPAC Name: 1,3-bis(bromomethyl)benzene
Synonyms: benzene, 1,3-bis(bromomethyl)-
Molecular Weight (g/mol): 263.96
Molecular Formula: C8H8Br2
InChi Key: OXHOPZLBSSTTBU-UHFFFAOYSA-N
SMILES: BrCC1=CC(CBr)=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (7)
  • (2)
  • (7)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)