accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 626-17-5

IUPAC Name: benzene-1,3-dicarbonitrile
Synonyms: isophthalonitrile dinitrile of isophthalic acid isoftalonitril isophthalodinitrile nitril kyseliny isoftalove isoftalodinitril 1,3-benzendikarbonitril m-phthalodinitrile 1,3-benzodinitrile
Molecular Weight (g/mol): 128.13
Molecular Formula: C8H4N2
InChi Key: LAQPNDIUHRHNCV-UHFFFAOYSA-N
SMILES: N#CC1=CC(=CC=C1)C#N
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)