accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CH2N(E/Z)

CAS RN 626-34-6

IUPAC Name: ethyl 3-aminobut-2-enoate
Synonyms: ethyl 3-aminobut-2-enoate
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: YPMPTULBFPFSEQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C=C(C)N
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)