accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrBr

CAS RN 626-39-1

IUPAC Name: 1,3,5-tribromobenzene
Synonyms: 1,3,5-tribromobenzene
Molecular Weight (g/mol): 314.8
Molecular Formula: C6H3Br3
InChi Key: YWDUZLFWHVQCHY-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=CC(Br)=C1
Boiling Point 
  • (8)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (8)
  • (2)
  • (2)
  • (8)
  • (3)
  • (7)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)