accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 626-77-7

IUPAC Name: propyl hexanoate
Synonyms: propyl hexanoate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: HTUIWRWYYVBCFT-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results