accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OH

CAS RN 626-89-1

IUPAC Name: 4-methylpentan-1-ol
Synonyms: 4-methyl-1-pentanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: PCWGTDULNUVNBN-UHFFFAOYSA-N
SMILES: CC(C)CCCO
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)