accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2O

CAS RN 626-97-1

IUPAC Name: pentanamide
Synonyms: pentanamide
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: IPWFJLQDVFKJDU-UHFFFAOYSA-N
SMILES: CCCCC(N)=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)