accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HO

CAS RN 6261-22-9

IUPAC Name: pent-2-yn-1-ol
Synonyms: pent-2-yn-1-ol
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: WLPYSOCRPHTIDZ-UHFFFAOYSA-N
SMILES: CCC#CCO
Boiling Point 
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)