CAS RN 62610-50-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals Agarose Beads 6% B-CL (Cross-linked)
CAS: 62610-50-8