accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 626207-32-7

IUPAC Name: bromozincylium; nonan-1-ide
Synonyms: bromozincylium; nonan-1-ide
Molecular Weight (g/mol): 272.54
Molecular Formula: C9H19BrZn
InChi Key: OXSAWWPTOCNGOR-UHFFFAOYSA-M
SMILES: [Zn+]Br.CCCCCCCC[CH2-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results