accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSH

CAS RN 6263-65-6

IUPAC Name: 1-phenylethane-1-thiol
Synonyms: 1-phenylethanethiol
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: QZZBJCFNHPYNKO-UHFFFAOYNA-N
SMILES: CC(S)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)