accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6263-83-8

IUPAC Name: 1,5-diphenylpentane-1,5-dione
Synonyms: 1,5-diphenylpentane-1,5-dione
Molecular Weight (g/mol): 252.31
Molecular Formula: C17H16O2
InChi Key: YOLLTWVIOASMFW-UHFFFAOYSA-N
SMILES: O=C(CCCC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results