accessibility menu, dialog, popup

UserName

Viewing profile as user:
SONNNNN

CAS RN 62645-28-7

IUPAC Name: 6-(benzenesulfinyl)-[1,2,3,4]tetrazolo[1,5-b]pyridazine
Synonyms: 6-(benzenesulfinyl)-[1,2,3,4]tetrazolo[1,5-b]pyridazine
Molecular Weight (g/mol): 245.26
Molecular Formula: C10H7N5OS
InChi Key: JFSXSNSCPNFCDM-UHFFFAOYNA-N
SMILES: O=S(C1=CC=CC=C1)C1=NN2N=NN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results