accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6265-92-5

IUPAC Name: 2-(4-methoxyphenyl)-1,3-benzothiazole
Synonyms: 2-(4-methoxyphenyl)-1,3-benzothiazole
Molecular Weight (g/mol): 241.31
Molecular Formula: C14H11NOS
InChi Key: AOPZIJQISHFZBN-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results