CAS RN 6267-02-3
IUPAC Name:
9,9-dimethyl-9,10-dihydroacridine
Synonyms:
9,9-dimethyl-10H-acridine
Molecular Weight (g/mol):
209.29
Molecular Formula:
C15H15N
InChi Key:
JSEQNGYLWKBMJI-UHFFFAOYSA-N
SMILES:
CC1(C)C2=CC=CC=C2NC2=CC=CC=C12
1
–
1
of
1
results
1
9,10-Dihydro-9,9-dimethylacridine 98.0+%, TCI America™
CAS: 6267-02-3 Molecular Formula: C15H15N Molecular Weight (g/mol): 209.29 MDL Number: MFCD00030130 InChI Key: JSEQNGYLWKBMJI-UHFFFAOYSA-N Synonym: 9,9-Dimethyl-9,10-dihydroacridine PubChem CID: 22647 IUPAC Name: 9,9-dimethyl-10H-acridine SMILES: CC1(C2=CC=CC=C2NC3=CC=CC=C31)C