accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62673-31-8

IUPAC Name: bromozincylium; phenylmethanide
Synonyms: bromozincylium; phenylmethanide
Molecular Weight (g/mol): 236.42
Molecular Formula: C7H7BrZn
InChi Key: WRSWIWOVJBYZAW-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)