accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 627-37-2

IUPAC Name: methyl(prop-2-en-1-yl)amine
Synonyms: N-methylallylamine
Molecular Weight (g/mol): 71.12
Molecular Formula: C4H9N
InChi Key: IOXXVNYDGIXMIP-UHFFFAOYSA-N
SMILES: CNCC=C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)