accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3OH

CAS RN 627-59-8

IUPAC Name: 5-methylhexan-2-ol
Synonyms: 5-methyl-2-hexanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: ZDVJGWXFXGJSIU-UHFFFAOYNA-N
SMILES: CC(C)CCC(C)O
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)