CAS RN 627-76-9
CAS RN 627-76-9
IUPAC Name:
2-aminoheptanedioic acid
Synonyms:
2-aminopimelic acid
Molecular Weight (g/mol):
175.18
Molecular Formula:
C7H13NO4
InChi Key:
JUQLUIFNNFIIKC-UHFFFAOYNA-N
SMILES:
NC(CCCCC(O)=O)C(O)=O
1
–
1
of
1
results
1
DL-2-Aminopimelic Acid 98.0+%, TCI America™
CAS: 627-76-9 Molecular Formula: C7H13NO4 Molecular Weight (g/mol): 175.184 MDL Number: MFCD00053314 InChI Key: JUQLUIFNNFIIKC-UHFFFAOYSA-N PubChem CID: 101122 ChEBI: CHEBI:64305 IUPAC Name: 2-aminoheptanedioic acid SMILES: C(CCC(=O)O)CC(C(=O)O)N