accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NOHO

CAS RN 627-95-2

IUPAC Name: hydrogen 5-aminopentanoic acid chloride
Synonyms: hydrogen 5-aminovaleric acid chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: BLOIUFYKQCCAGP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCCCC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)