accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6270-57-1

IUPAC Name: (2Z,5Z)-bis[ethoxy(hydroxy)methylidene]oxolane-3,4-dione
Synonyms: (2Z,5Z)-bis[ethoxy(hydroxy)methylidene]oxolane-3,4-dione
Molecular Weight (g/mol): 244.2
Molecular Formula: C10H12O7
InChi Key: YCNONGXDPIGWCP-XOHWUJONSA-N
SMILES: CCO\C(O)=C1/O\C(=C(\O)OCC)C(=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results