accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 62716-65-8

IUPAC Name: 4-ethoxyphenyl 4-butylbenzoate
Synonyms: 4-ethoxyphenyl 4-butylbenzoate
Molecular Weight (g/mol): 298.38
Molecular Formula: C19H22O3
InChi Key: DDDWTNWRNNRWNQ-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C(=O)OC1=CC=C(OCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results