accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6274-12-0

IUPAC Name: diethylazanium bromide
Synonyms: diethylamine, hydrobromide
Molecular Weight (g/mol): 154.05
Molecular Formula: C4H12BrN
InChi Key: AATGHKSFEUVOPF-UHFFFAOYSA-N
SMILES: [Br-].CC[NH2+]CC
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results