accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6276-54-6

IUPAC Name: hydrogen 3-chloropropan-1-amine chloride
Synonyms: hydrogen 3-chloropropan-1-amine chloride
Molecular Weight (g/mol): 130.01
Molecular Formula: C3H9Cl2N
InChi Key: IHPRVZKJZGXTBQ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCCCl
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (6)
  • (5)
  • (5)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)